Acro® 50 Vent Filters, Cytiva (Formerly Pall Lab)
Fournisseur: Cytiva
New Product
Certificats
À propos de cet article
Reusable, in-line filters with 50 mm hydrophobic PTFE membranes in polypropylene housing.
- Reusable
- Hydrophobic
- Polypropylene housing
Large surface area provides greater throughput. Ideal for venting bioreactors, fermentation tanks, and carboys. Also for filtration of aggressive solvents and sterile gas purging of culture vessels. Stepped hose-barb connectors fit 6.4 to 12.7 mm (¹/₄ to ¹/₂") I.D. tubing. Autoclavable at 121 °C (250 °F) for 30 minutes.
Ordering information: All are individually packaged, except for 28144-111, which comes in six bags of 12.
Spécifications
- Catalog No:
- CA28143-955
- CA28143-718
- CA28143-616
- CA89201-642
- CA28143-558
- CA28144-111
- CA28143-966
- CA16003-654
- Effective filtration area:
- 19.6 cm²
- 19.6 cm²
- 19.6 cm²
- 19.6 cm²
- 19.6 cm²
- 19.6 cm²
- 19.6 cm²
- 19.6 cm²
- Max. ambient temperature:
- 130 °C (266 °F)
- 130 °C (266 °F)
- 130 °C (266 °F)
- 130 °C (266 °F)
- 130 °C (266 °F)
- 130 °C (266 °F)
- 130 °C (266 °F)
- 130 °C (266 °F)
- Max. operating pressure:
- 4.1 bar (60 psi) at ambient temperature
- 4.1 bar (60 psi) at ambient temperature
- 4.1 bar (60 psi) at ambient temperature
- 4.1 bar (60 psi) at ambient temperature
- 4.1 bar (60 psi) at ambient temperature
- 4.1 bar (60 psi) at ambient temperature
- 4.1 bar (60 psi) at ambient temperature
- 4.1 bar (60 psi) at ambient temperature
- Overall length:
- 8.2 cm (37/32")
- 8.2 cm (37/32")
- 8.2 cm (37/32")
- 8.2 cm (37/32")
- 8.2 cm (37/32")
- 8.2 cm (37/32")
- 8.2 cm (37/32")
- 8.2 cm (37/32")
- Ø:
- 7.3 cm (27/8")
- 7.3 cm (27/8")
- 7.3 cm (27/8")
- 7.3 cm (27/8")
- 7.3 cm (27/8")
- 7.3 cm (27/8")
- 7.3 cm (27/8")
- 7.3 cm (27/8")