VWR® Lab Wrap, Clear Plastic
VWR® clear plastic lab wrap, roll core 4.2 cm, 30.5 m (100'), 305 mm (12")
Catalog # 46610-056
Supplier: VWR International
Jump to:
Product Details & Documents
Designed specifically for the lab, clear 100% polyethylene wrap handles easily without tangling, yet seals tightly to a beaker, flask, or Petri dish.
- Low tangle properties
- High cling properties
- Ultra-clean film
Unique formulation yields ultra-clean film to ensure sample purity and eliminate contamination. Dispenser carton features a metal tear-off bar as well as flaps designed to keep the roll in the box.
Specifications
- Largeur:305 mm (12")
- Désignation:VWR® clear plastic lab wrap, roll core 4.2 cm
- Longueur:30.5 m (100')