Metrigard® Filters, Cytiva (Formerly Pall Lab)
Catalog # CA28150-371
Supplier: Cytiva
Specifications
- Ø:4.7 cm
- Particle retention:0.5 µm
- Flow rate:Medium
- Cat. no.:CA28150-371
Specifications
About this item
This depth-type filter medium is useful for prolonging membrane filter life in liquid systems that contain substantial amounts of particulate matter.
- Depth-type
- Finer glass fibers
Made of finer glass fiber than standard prefilters, with 5% acrylic binder. Liquid nominal rating: 0.5 µm.