Glass Fiber Filters, Extra Thick, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Certificates
About this item
High contaminant-holding capacity makes these filters efficient as depth filters and useful for prefiltration of viscous biological solutions.
- High capacity
- Useful for prefiltration
- Thicker than type A/E
Three times thicker than type A/E filters, with greater contaminant holding capacity. Useful for prefiltration of viscous biological solutions. Contain 5% acrylic binder for additional wet strength.
Specifications
- Catalog No:
- CA28150-779
- CA28150-735
- CA28150-713
- Particle retention:
- 1.0 µm
- 1.0 µm
- 1.0 µm
- Flow rate:
- Medium-Fast
- Medium-Fast
- Medium-Fast