Binder-Free Glass Fiber Filters, Cytiva (Formerly Pall Lab)
: Cytiva
Superior grade glass fiber filters for a variety of air sampling and liquid filtration methods.
- Superior grade
- For a variety of applications
Type A/B is designed for diagnostic applications, gravimetric analysis, and sample filtration. Use Type A/C for cell harvesting, prefiltration, and solids testing. Type A/D makes an excellent prefilter for solutions with a heavy load of large-sized particulate. Manufactured with high-quality borosilicate glass microfibers without binder for assurance of purity. Excellent wet strength for ease of handling and filter integrity.
: All three types are also available as 20.3×25.4 cm (8×10") sheets.
- Catalog No:
- CA28150-976
- CA28150-975
- CA28150-984
- CA28150-938
- CA28150-981
- CA28150-999
- CA28150-979
- CA28150-996
- CA28150-978
- CA28150-995
- Grade:
- A/B
- A/B
- A/C
- A/B
- A/C
- A/D
- A/C
- A/D
- A/B
- A/D
- Thickness:
- 660 µm
- 660 µm
- 279 µm
- 660 µm
- 279 µm
- 686 µm
- 279 µm
- 686 µm
- 660 µm
- 686 µm
- Particle retention:
- 1.0 µm
- 1.0 µm
- 1.0 µm
- 1.0 µm
- 1.0 µm
- 3.1 µm
- 1.0 µm
- 3.1 µm
- 1.0 µm
- 3.1 µm
- Flow rate:
- Fast
- Fast
- Fast
- Fast
- Fast
- Very Fast
- Fast
- Very Fast
- Fast
- Very Fast
Frequently Bought Together