TCLP Glass Fiber Filters, Cytiva (Formerly Pall Lab)
: Cytiva
TCLP glass fiber filters are designed to meet requirements for use in U.S. EPA SW-846 Method 1311: Toxic Characteristics Leaching Procedure (TCLP).
- Reduces filtration costs and premature clogging when filtering difficult-to-filter or highly contaminated solutions
- Extends filter life and reduces final filter changes with high capacity prefilters
- Eliminates sample contamination, binder-free borosilicate glass fiber has no added extractables
Filters a wide range of particulate loads and viscous solutions with a selection of filter thicknesses from which to choose.
- Catalog No:
- CA29900-008
- CA28149-457
- CA28149-456
- CA28149-455
- Particle retention:
- 0.7 µm
- 0.7 µm
- 0.7 µm
- 0.7 µm
- Flow rate:
- Medium
- Medium
- Medium
- Medium
Frequently Bought Together