VWR® Qualitative Crepe Fluted Circles, Folded, Grade 315
Supplier: VWR International
Certificates
About this item
Crepe fluted circles are ideal for gelatinous and coarse precipitates.
- Coarse porosity
- Fast flow rate
- Assorted sizes
Specifications
- Catalog No:
- CA28331-081
- CA28331-060
- CA28331-048
- CA28331-026
- CA28331-128
- CA28331-106
- Particle retention:
- 25 µm
- 25 µm
- 25 µm
- 25 µm
- 25 µm
- 25 µm
- Porosity:
- Coarse
- Coarse
- Coarse
- Coarse
- Coarse
- Coarse
- Flow rate:
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast