VWR® Qualitative Filter Paper, Grade 410
Supplier: VWR International
Certificates
About this item
Grade 410 filter paper is useful in analytical operations involving filtration of cuprous oxide, lead sulfate, barium sulfate, manganese carbonate, and more.
Specifications
- Catalog No:
- CA28297-826
- CA28321-077
- CA28321-135
- CA28321-113
- Particle retention:
- 1 µm
- 1 µm
- 1 µm
- 1 µm
- Porosity:
- Fine
- Fine
- Fine
- Fine
- Flow rate:
- Slow
- Slow
- Slow
- Slow