ContactUsLinkComponent
Multi-Well Plate Vacuum Manifold, Cytiva (Formerly Pall Lab)
Catalog # CA28148-486
Supplier:  Cytiva
undefined
Multi-Well Plate Vacuum Manifold, Cytiva (Formerly Pall Lab)
Catalog # CA28148-486
Supplier:  Cytiva
Supplier Number:  5017
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Multi-Well Plate Vacuum Manifold
  • Cat. no.:
    CA28148-486
  • Supplier No.:
    5017
Specifications
About this item
Optimized for the vacuum filtration of AcroPrep™ and AcroWell™ multi-well plates, manifold can also be used with other multi-well filter plates that meet ANSI/SBS X-2004 specifications.
  • For vacuum filtration
  • Can be used with other multi-well plates
  • Anodized aluminum
  • Adapter collar holds filter plates tightly to receiver plates for centrifugation
Manifold is constructed of anodized aluminum and is designed to facilitate the filtration of liquid through a multi-well filter plate into a receiver plate for collection.
Vacuum manifold unit includes a Delrins spacer block designed to accommodate standard 350 μL receiver plates. The spacer block has been optimized to reduce the space between the receiver plate and the filter plate during vacuum filtration. Optional spacer block available for use with 1 mL receiver plates.
Ordering information: Manifold includes O-ring, gasket, and spacer block. Control block on manifold includes vacuum pressure gauge, metering valve, release valve, and 6.4mm (1/4") hose barb connection for vacuum line attachment. Replacement accessory kit includes O-ring, gasket, and allen wrench.