StatSpin® Microhematocrit Tubes, Iris Sample Processing
À propos de cet article
These microhematocrit tubes use capillary action to draw samples quickly.
- Capillary Action Draws Samples Quickly
- Material: Non-Breakable Plastic for Safety
- Heparin Tubes for Finger-Stick Samples
- Untreated Tubes for Anti-Coagulated Venous Samples
- Available Lengths: 75 mm & 40 mm
Tubes are available with heparin for finger-stick samples or untreated for anti-coagulated venous samples
SafeCrit® hematocrit tubes are made of non-breakable, 100% plastic to eliminate the potential safety hazards of broken glass.
The 40 mm tubes are made for use with the MP multipurpose centrifuge (87002-248 series) and the CritSpin™ microhematocrit centrifuge (87002-206 series). The 75 mm tubes are made for use in conventional centrifuges that support the full length of the tube.
6 Options Available Below
Product Details & Documents
These microhematocrit tubes use capillary action to draw samples quickly.
- Capillary Action Draws Samples Quickly
- Material: Non-Breakable Plastic for Safety
- Heparin Tubes for Finger-Stick Samples
- Untreated Tubes for Anti-Coagulated Venous Samples
- Available Lengths: 75 mm & 40 mm
Tubes are available with heparin for finger-stick samples or untreated for anti-coagulated venous samples
SafeCrit® hematocrit tubes are made of non-breakable, 100% plastic to eliminate the potential safety hazards of broken glass.
The 40 mm tubes are made for use with the MP multipurpose centrifuge (87002-248 series) and the CritSpin™ microhematocrit centrifuge (87002-206 series). The 75 mm tubes are made for use in conventional centrifuges that support the full length of the tube.
Product Family Options
Informations sur le produit
- DésignationLongueurVolumeAvailabilityPrice
- SafeCrit plastic capillary tube, Sodium Heparin anticoagulant40 mm20 µlPack of 100$58.32Case of 1000$583.20Supplier Number: HP8H-10
- SafeCrit plastic capillary tube, Sodium Heparin anticoagulant75 mm40 µlPack of 100 (1/Itm/Bx)$42.04Case of 1000 (1/ Itm/Bx)$420.40Supplier Number: HP4H-10
- SafeCrit plastic capillary tube, no anticoagulant75 mm40 µlPack of 100 (1/Itm/Bx)$46.85Case of 1000 (1/ Itm/Bx)$468.50Supplier Number: HP4U-10
- Glass capillary tube, Ammonium Heparin anticoagulant40 mm9 µlPack of 100$34.88Case of 1000$348.80Supplier Number: HT9H-10
- Glass capillary tube, no anticoagulant40 mm9 µlPack of 100$30.88Case of 1000$308.80Supplier Number: HT9U-10
- SafeCrit plastic capillary tube, no anticoagulant40 mm20 µlPack of 100$54.50Case of 1000$545.05Supplier Number: HP8U-10