SAF-T-FIT™ Plus N1115 Disposable Respirators, Honeywell Safety
Supplier: Honeywell Safety Products
Certificates
About this item
Ideal for medical, grinding, polishing, sanding, woodworking, and sweeping applications or other dusty operations.
- Offer latex-free N95 protection from particulate aerosols free of oil
- Adjustable boomerang nose seal fits securely and comfortably
- Color-coded for quick size identification
- NIOSH 95 approved
For use with particulate aerosols free of oil. Respirators feature an adjustable boomerang nose seal for a secure fit and added comfort. Nose seal is color-coded for easy size recognition.
Frequently Bought Together