Segregating hazardous solid waste from normal waste using bench top solid waste containers, helps to keep lab RCRA compliant.
- It has a small footprint and can easily sit in the corner of a fume hood or bench
- Made of chemically resistant HDPE with a detachable base
Convenient way to discard of contaminated solid wastes right where you work. Not carrying contaminated items across a lab to a larger waste container means preventing cross contamination. The latching lid reduces solvent emissions from the waste inside, keeping the air you breathe safer and healthier.
Comes with detachable base and lid with gasket is an ideal containment system for contaminated solid wastes, preventing possible cross contamination of products. The latching lid significantly reduces the emission of solvent-soaked solid waste back into the workspace. ideal if you routinely handle radioisotopes, beta-emitters, wet silica gel, contaminated gloves, kim-wipes, filter papers, wiping rags, pipettes, pipette bulbs, and other disposables.
Footprint: 13×13" (with base), 7×7" (without base).
Option are available for solid waste containers with red or yellow lids comes with a single sample liner bag. Additional can liners are available in packs of 100 or 250, red or yellow or biohazard bags.
Poly liner bags: 45.72 cm (18") length, 35.56 cm (14") width, 0.05 mm (thick), 5.4 lbs./100 and 8.5 lbs./250 (weight).