PUREZERO* HG3 cream latex gloves are designed specifically to meet the needs of semi-conductor electronics medical device and other cleanroom manufacturing applications.
- Recommended for use in ISO Class 3 or higher and Grade B/C/D cleanrooms
- Maximum Particles 0.5 µm/cm² less than 1200
- 30.2 cm (12") length with beaded cuff available in hand-specific (sterile) and ambidextrous (non-sterile) options
- AQL 1.5 for pinholes
- Silicone and powder free
- Tested against 35 chemicals 14 chemotherapy drugs and fentanyl
- Recommended for applications such as semi-conductor electronics medical device and other cleanroom manufacturing applications
- Fingertip Thickness: 0.20 mm/7.9 mil and palm thickness 0.18 mm/7.1 mil
- Textured fingertips with smooth finish
PUREZERO* HG3 Cream Latex Cleanroom Gloves are designed for semiconductor pharmaceutical and medical device manufacturing applications. These gloves are clean processed (washed repeatedly in deionized water) to ensure consistent control of low particles and extractables and are recommended for use in ISO Class 3 or higher and Grade B/C/D cleanrooms.
These gloves are available in both non sterile ambidextrous and sterile hand specific options. The gloves are 12" long with a smooth finish and feature a beaded cuff with a 7.9 mil fingertip thickness and textured fingertips and palms. They have triple layer cleanroom compatible packaging (double poly bagged with poly case liner).
- Catalog No:
- 77681-960
- 77681-958
- 77681-964
- 77681-948
- 77681-946
- 77681-952
- 77681-950
- 77681-956
- 77681-954
- 77681-966
- 77681-970
- 77681-968
- 77681-962
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Hand-specific
- Ambidextrous
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Natural rubber latex
- Natural rubber latex
- Natural rubber latex
- Natural rubber latex
- Natural rubber latex
- Natural rubber latex
- Natural rubber latex
- Natural rubber latex
- Natural rubber latex
- Natural rubber latex
- Natural rubber latex
- Natural rubber latex
- Natural rubber latex
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Finger thickness:
- 7.9 mil
- 7.9 mil
- 7.9 mil
- 7.9 mil
- 7.9 mil
- 7.9 mil
- 7.9 mil
- 7.9 mil
- 7.9 mil
- 7.9 mil
- 7.9 mil
- 7.9 mil
- 7.9 mil
- Palm thickness:
- 7.1 mil
- 7.1 mil
- 7.1 mil
- 7.1 mil
- 7.1 mil
- 7.1 mil
- 7.1 mil
- 7.1 mil
- 7.1 mil
- 7.1 mil
- 7.1 mil
- 7.1 mil
- 7.1 mil
- Sterility:
- Sterile
- Sterile
- Sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Sterile
- Non sterile
- Sterile
- Sterile
- Sterile
- Sterile