Buffer Solutions
Buffer Solutions
 :   Ohaus
Buffer Solutions
 :  Ohaus
Buffer Solutions
Buffer Solutions
Buffer solution, 250 ml bottle.
Product Family Options
  • Density
    pH
  • Buffer solution
    1.68
    Each
    Each - $53.57
    $53.57
    Each
       $53.57
     : 30100424
  • Buffer solution
    4.01
    Each
    Each - $46.90
    $46.90
    Each
       $46.90
     : 30100425
  • Buffer solution
    6.86
    Each
    Each - $46.90
    $46.90
    Each
       $46.90
     : 30100426
  • Buffer solution
    7.00
    Each
    Each - $46.90
    $46.90
    Each
       $46.90
     : 30100427
  • Buffer solution
    9.18
    Each
    Each - $46.90
    $46.90
    Each
       $46.90
     : 30100428
  • Buffer solution
    10.01
    Each
    Each - $46.90
    $46.90
    Each
       $46.90
     : 30100429
  • Buffer solution
    12.45
    Each
    Each - $53.57
    $53.57
    Each
       $53.57
     : 30100440