Anti-Cleaved-F13A1 (G39) Rabbit Polyclonal Antibody
Catalog # 77702-254
Supplier: AFG Bioscience
New Product
Specifications
- Form:Liquid
- Gene ID:P00488
- Antigen synonyms:Coagulation factor XIII|Transglutaminase. plasma|A1 polypeptide)|Coagulation factor XIII A1 subunit|Transglutaminase A chain|A subunit|plasma|Protein-glutamine gamma-glutamyltransferase A chain|Transglutaminase|Coagulation factor XIIIa|FSF|Coagulation factor XIII A1 polypeptide|bA525O21.1 (coagulation factor XIII|Fibrinoligase|F13A_HUMAN|F13a1|Protein glutamine gamma glutamyltransferase A chain|TGase|A polypeptide|Factor XIIIA|F13A|Coagulation factor XIII A chain|Fibrin stabilizing factor
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human Factor XIIIa.
- Purification:Antigen affinity purified
- Cat. no.:77702-254
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: Cleaved-F13A1 (G39)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human