Anti-CYB5A Rabbit Polyclonal Antibody
Fournisseur: AFG Bioscience
New Product
Certificats
À propos de cet article
ELISA:1:1000-1:2000, WB:1:200-1:1000, IHC:1:50-1:200
Type:
Antigen: CYB5A
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Spécifications
- Catalog No:
- 77714-574
- 77714-575
- Antigen synonyms:
- Cytochrome b5 type A (microsomal)|CYB5_HUMAN|MCB 5|Microsomal cytochrome b5 type A|Microsomal cytochrome b5|Cytochrome b 5|Cytochrome b5|Cyb5a|CYB 5|Cytochrome b5 (microsomal)|Cytochrome b5 type A|CYB5|CYB 5A|Type 1 cyt b5|MCB5
- Cytochrome b5 type A (microsomal)|CYB5_HUMAN|MCB 5|Microsomal cytochrome b5 type A|Microsomal cytochrome b5|Cytochrome b 5|Cytochrome b5|Cyb5a|CYB 5|Cytochrome b5 (microsomal)|Cytochrome b5 type A|CYB5|CYB 5A|Type 1 cyt b5|MCB5
- Gene ID:
- P00167
- P00167
- Immunogen:
- Fusion protein of Human CYB5A
- Fusion protein of Human CYB5A
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Spécifications
Frequently Bought Together