Anti-Cleaved-ITIH4 (R661) Rabbit Polyclonal Antibody
Catalogue # 77703-354
Fournisseur: AFG Bioscience
New Product
Spécifications
- Form:Liquid
- Gene ID:Q14624
- Antigen synonyms:ITIH4|ITIHL1|IHRP|OTTHUMP00000197121|PRO1851|Inter-alpha-trypsin inhibitor family heavy chain-related protein|35 kDa inter-alpha-trypsin inhibitor heavy chain H4|OTTHUMP00000213834|PK120|PK-120|inter-alpha-trypsin inhibitor, heavy chain-like, 1|ITI-HC4|OTTHUMP00000197120|OTTHUMP00000213869|Plasma kallikrein sensitive glycoprotein 120|Inter-alpha-inhibitor heavy chain 4|ITI heavy chain H4|H4P|gp120|ITIH4_HUMAN|Inter-alpha-trypsin inhibitor heavy chain H4
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human ITI-H4 70k.
- Purification:Antigen affinity purified
- Cat. no.:77703-354
- Packaging:Vial
Spécifications
À propos de cet article
IHC:1:100-1:300, ELISA:1:10000
Type:
Antigen: Cleaved-ITIH4 (R661)
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human