Anti-GRIN2D Rabbit Polyclonal Antibody
77702-713
New Product
- Antigen symbol:GRIN2D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:O15399
- Antigen synonyms:GluN2D|NR2D|Glutamate receptor ionotropic|NMDA 2D|Glutamate receptor, ionotropic, N-methyl D-aspartate 2D|NMDAR2D|NMDE4|Glutamate [NMDA] receptor subunit epsilon 4|EB11|N methyl D aspartate receptor subtype 2D|N methyl d aspartate receptor subunit 2D|Grin2d|N-methyl D-aspartate receptor subtype 2D|Glutamate [NMDA] receptor subunit epsilon-4|NMDE4_HUMAN|estrogen receptor binding CpG island|glutamate receptor ionotropic NMDA 2D
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human NMDAε4.
- Purification:Antigen affinity purified
- Cat. no.:77702-713
- Packaging:Vial
IHC:1:100-1:300, ELISA:1:40000
Type:
Antigen: GRIN2D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat