Anti-HIST1H2AB HEK293F cells Recombinant Monoclonal Antibody [clone: 3B12]
Catalog # 77691-274
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:HIST1H2AB
- Clonality:Recombinant Monoclonal
- Clone:3B12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:HEK293F cells
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P04908
- Antigen synonyms:AND|H2AFA|HIST1H2AB|Histone H2A/a|HIST1H2AE|Histone H2A.2|Histone H2A type 1-B/E|H2AFM|Histone H2A/m
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide
- Purification:Affinity purified
- Cat. no.:77691-274
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:2000
Type:
Antigen: HIST1H2AB
Clonality: Recombinant Monoclonal
Clone: 3B12
Conjugation: Unconjugated
Epitope:
Host: HEK293F cells
Isotype: Rabbit IgG
Reactivity: Human