Anti-HAMP HEK293F cells Recombinant Monoclonal Antibody [clone: 4C5]
Catalog # 77691-268
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:HAMP
- Clonality:Recombinant Monoclonal
- Clone:4C5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:HEK293F cells
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P81172
- Antigen synonyms:PLTR|Hepcidin|Hepc20|UNQ487/PRO1003|Liver-expressed antimicrobial peptide 1|Hepcidin-20|Hepcidin-25|Putative liver tumor regressor|HAMP|LEAP1|LEAP-1|Hepc25|HEPC
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:A synthesized peptide derived from Human HAMP
- Purification:Affinity purified
- Cat. no.:77691-268
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:5000, IHC:1:50-1:200
Type:
Antigen: HAMP
Clonality: Recombinant Monoclonal
Clone: 4C5
Conjugation: Unconjugated
Epitope:
Host: HEK293F cells
Isotype: Rabbit IgG
Reactivity: Human