Anti-Histone H1 (Phospho-Thr3) Rabbit Polyclonal Antibody
Catalog # 77698-452
Supplier: AFG Bioscience
New Product
Specifications
- Form:Liquid
- Gene ID:Q02539, P16403, P16402, P10412
- Antigen synonyms:Histone H1d|Histone H1.1|Histone H1c|H1F2|Histone H1.4|HIST1H1C|Histone H1s-1|Histone H1s-4|HIST1H1D|HIST1H1E|H1F3|Histone H1b|Histone H1a|H1F4|H1F1|Histone H1s-2|HIST1H1A|Histone H1.2|Histone H1.3
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthetic Peptide
- Purification:Affinity purified
- Cat. no.:77698-452
- Packaging:Vial
Specifications
About this item
WB:1:1000-2000
Type:
Antigen: Histone H1 (phospho-Thr3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat