Anti-BIN1 Rabbit Polyclonal Antibody
100 µg, Vial
77701-151
: A17403-100UG
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: BIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Taille:100 µg
- Packaging:Vial
- Western blot:Yes
- Clonality:Polyclonal
- Purification:Antigen affinity purified
- Form:Liquid
- Isotype:IgG
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Host:Rabbit
- Shipping temperature:Shipped on blue ice
- Cat. No.:77701-151
- Reactivity:Human|Rat|Mouse
- Gene ID:O00499
- Antigen synonyms:Myc box-dependent-interacting protein 1|amphiphysin-like|Amphiphysin-like protein|Amphiphysin II|MGC105358|Bridging integrator 1|Amphiphysin like protein|AMPH2|Box Dependant MYC Interacting Protein 1|BIN1_HUMAN|Amphiphysin 2|AMPHL|SH3P9|DKFZp547F068|MGC10367|Myc box dependent interacting protein 1|Bin1|AMPH 2|Box-dependent myc-interacting protein 1
- Immunogen:Synthesized peptide derived from the C-terminal region of Human Amphiphysin II
- Conjugation:Unconjugated
- Antigen symbol:BIN1
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- ELISA:Yes



