PinPort™ for Catheters and Tubing
Supplier: INSTECH LABORATORIES, INC
New Product
Certificates
About this item
The PinPort™ provides fast, aseptic access to externalized rodent catheters or tubing for dosing or blood sampling. It can reduce study costs and improve compliance with the 3Rs.
- Fast access
- Closed system for improved patency
- Needle free
- Low cost
- Manufactured in USA
- Sterilized by ETO gas
The system consists of a lightweight port and a mating injector which can access the port hundreds of times.
Quickly infuse or withdraw from a catheter in a fraction of the time it takes with plugs and blunt needles. The PinPort™ is a closed system that permits aseptic technique. This can reduce infection, improve patency, and ultimately reduce the number of animals required for a study. To ensure safety, the injector is recessed and provides complete needle stick protection for personnel.
Caution: For research use only.
Frequently Bought Together