Luer Stubs (Blunt Needles)
Fournisseur: INSTECH LABORATORIES, INC
New Product
Certificats
À propos de cet article
Use these female luer stubs to connect infusion tubing or catheters to syringes or other male Luer connectors. The hubs are polypropylene and the tips are stainless steel. The tips are deburred to prevent damage to tubing.
- Female Luers
- Polypropylene hubs
- Stainless steel tips
- Deburred tips
Blunt tip needles (Luer stub adaptors) have numerous laboratory applications. When dosing, sampling, or infusing, these blunt needles ensure a secure and reliable connection to in-dwelling catheters. They provide a smooth, burr-free needle edge which prevents inadvertent damage to the lumen of the catheter caused by inferior needles. These high-quality, low-cost blunt needles are a critical tool for dosing, sampling, flushing, and locking catheters.
Frequently Bought Together