Aluminum Weighing Dishes, Oil Free, Heathrow Scientific
Supplier: Heathrow Scientific
New Product
Certificates
About this item
Oil free dishes for general-purpose weighing, dispensing, storage, food processing, evaporating or drying.
- Processed and packaged in clean room environment
- Lot traceable
Passed through a rigorous, three-step cleansing process to remove forming oil or residue for more reliable weighing and moisture sample analysis.
Frequently Bought Together