Saturix C1E Enhanced Polyester/Cellulose Wipes
Supplier:   FG CLEAN WIPES TE
Saturix C1E Enhanced Polyester/Cellulose Wipes
Supplier:  FG CLEAN WIPES TE
Certificates
Saturix C1E Enhanced Polyester/Cellulose Wipes
Saturix C1E Enhanced Polyester/Cellulose Wipes
3 Options Available Below
Product Details & Documents
These cleanroom wipes combine the versatility of a polyester/cellulose substrate with added strength, softness, and enhanced cleanliness. A special treatment further reduces ions, NVRs, and particulates, creating the highest level of cleanliness for a lint-free cleanroom wipe in its class.
  • 55% cellulose/45% polyester
  • Treated to remove impurities
  • Ultra-low extractables and ion levels for a polyester/cellulose non-woven wipe
  • Significantly lower in particles than other non-woven wipe fabrics
  • Pure and clean; chemical residue-free
  • Ideal for picking up aqueous spills
  • All the advantages of a best-in-class poly/cell wipe with critical cleanliness
FG Cleanwipes CE1 products are designed to meet the rigorous demands of cleanroom environments, offering enhanced strength, softness, and cleanliness through their polyester/cellulose blend. Treated to reduce ions, non-volatile residues (NVRs), and particulates, they provide superior lint-free performance that is ideal for contamination-sensitive applications.
Their versatility makes them effective for cleaning critical surfaces in controlled environments. Offering enhanced cleanliness, their advanced treatment minimizes particulate release, ensuring optimal cleanliness. Their design combines strength and durability with softness for precision cleaning without damage.
Targeted industries and customers:
Pharmaceutical and Biotech: Critical for sterile manufacturing and cleanroom maintenance.
Electronics: Ensures contamination-free environments for semiconductor and electronics assembly.
Medical device manufacturing: Supports precision cleaning in compliance with stringent cleanliness standards.
These wipes are an essential tool for industries that prioritize contamination control and process efficiency.
Product Family Options
Informations sur le produit
  • Matière
    Color
    Packaging
    W×L
    Availability
    Price
  • Polyester/cellulose
    White
    1200/bag, 12 bags/case
    4×4"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 14,400
    Case of 14,400 - $1,500.14
    $1,500.14
    Case of 14,400
       $1,500.14
    Supplier Number: 7-C1E-44L-00
  • Polyester/cellulose
    White
    300/bag, 12 bags/case
    9×9"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 3,600
    Case of 3,600 - $963.13
    $963.13
    Case of 3,600
       $963.13
    Supplier Number: 7-C1E-99L-00
  • Polyester/cellulose
    White
    150/bag, 18 bags/case
    12×12"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 2,700
    Case of 2,700 - $1,439.26
    $1,439.26
    Case of 2,700
       $1,439.26
    Supplier Number: 7-C1E-BBL-00