VWR® UV Transilluminators
VWR® UV Transilluminators
Supplier:   VWR International
VWR® UV Transilluminators
Supplier:  VWR International
Certificates
VWR® UV Transilluminators
VWR® UV Transilluminators
2 Options Available Below
Product Details & Documents
The VWR® UV transilluminator offers high-performance gel viewing in a modern, compact housing.
  • Hinged, UV-blocking safety cover
  • Scratch-resistant viewing surface
  • Adjustable UV intensity
The VWR® UV transilluminator features a large, scratch-resistant viewing surface that is evenly illuminated by four high-powered 14 W UV tubes. UV intensity is fully adjustable using the control knob. The long-lasting filter glass ensures a 302 nm mid-range output ideal for gels stained with ethidium bromide, SYBR green, and red stains. Models are available with either 115 or 230 V input.
Product Family Options
115 V
Informations sur le produit
  • Désignation
    Longueur d'onde
    Electrical
    Dimensions
    Availability
    Price
  • VWR® UV transilluminator
    302 nm
    115 V
    21×26 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,219.11
    $2,219.11
    Each
       $2,219.11
230 V
Informations sur le produit
  • Désignation
    Longueur d'onde
    Electrical
    Dimensions
    Availability
    Price
  • VWR® UV transilluminator
    302 nm
    230 V
    21×26 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,219.11
    $2,219.11
    Each
       $2,219.11