VWR® 900 Portable Conductivity Meters
New Product
The VWR-EC900 meter provide accurate and reliable measurements of conductivity, resistivity, TDS, salinity and temperature in any environment.
- High resolution color touch display (4.3")
- Five-point calibration with standard recognition
- IP65 waterproof
- Handheld
- Automatic or manual temperature compensation capabilities
- Auto-read, timed-read, and continuous-read measurement options
- Ability to save 1000 data sets for each parameter (GLP-compliant)
- Rechargeable battery
- Settable parameters, including cell constant, temperature compensation coefficient, and TDS factor
The easy to use VWR-EC900 portable meters provide accurate conductivity measurements inside and outside the lab. The intelligent operation system provides features including user managment, calibration management, electrode management, method management, and many other advanced capabilities.
: Three-year meter warranty. For application specific recommendations, please reference 'VWR® Electrode Selection Guide' located in the Related Information section.
: All meters include carrying case, electrode holders (2), meter silicone rubber case, touch screen pen, and hand strap.
Frequently Bought Together