ContactUsLinkComponent
VWR® Traceable® Pens for Conductivity and Dissolved Solids
VWR® Traceable® Pens for Conductivity and Dissolved Solids
Supplier:  VWR International
Certificates
undefined
VWR® Traceable® Pens for Conductivity and Dissolved Solids
Supplier:  VWR International
Specifications
  • Catalog No:
  • 89094-764
  • 89094-762
  • Model:
  • Dissolved solids meter
  • Conductivity meter
  • Conductivity range:
  • 0 - 1999 microsiemens, 0 - 19.99 millisiemens
  • 0 - 1999 microsiemens, 0 - 19.99 millisiemens
  • Conductivity resolution:
  • 1 microsiemen, 0.01 millisiemen
  • 1 microsiemen, 0.01 millisiemen
  • Conductivity accuracy:
  • ±1% full scale + 1 digit
  • ±1% full scale + 1 digit
  • TDS range:
  • 0 - 1999 ppm, 0 - 19.99 ppt
  • TDS resolution:
  • 1 ppm
  • TDS accuracy:
  • ±1% full scale + 1 digit
  • TDS conversion factor:
  • 0.4 - 1
  • Temperature range:
  • 0...50 °C (32...122 °F)
  • Temperature resolution:
  • 0.1°
  • Temperature accuracy:
  • ±0.5 °C (±1 °F)
  • ±0.5 °C (±1 °F)
  • Temperature compensation:
  • 20° and 25° (automatic)
  • 20° and 25° (automatic)
  • Temperature coefficient:
  • 0 - 4% per °C
  • 0 - 4% per °C
  • Dimensions:
  • 16.5×3.2×3.2 cm (6¹/₂×1¹/₄×1¹/₄")
  • 16.5×3.2×3.2 cm (6¹/₂×1¹/₄×1¹/₄")
  • Weight:
  • 63.8 g (2¹/₄ oz.)
  • 63.8 g (2¹/₄ oz.)