REDISHIP Auxiliary Air Chambers, Kewaunee®
Catalogue # CF5-1372-03
Fournisseur: Kewaunee
Product available on GSA Advantage®
Spécifications
- Color:Light Neutral
- Cat. no.:CF5-1372-03
- Shipping Weight:143 lbs.
- For Use With:6' Hoods
- Static Pressure:0.15" (730 cfm) @ 80 fpm, 0.25" (900 cfm) @ 100 fpm, 0.30" (1090 cfm) @ 120 fpm
- Supplier No.:CF5-1372-03
Spécifications
À propos de cet article
These chambers can be added to the top of any Supreme Air fume hood to convert it to an auxiliary air fume hood.
- Includes all necessary parts except a fan and ductwork
- Requires no modifications to the fume hood
- Chambers are made of cold-rolled steel
- Finish is light neutral color or snow white
Adding the chamber allows the fume hood to operate on up to 70% auxiliary air, saving expensive, conditioned room air from being exhausted.
Ordering information: REDISHIP units can be shipped within 48 hours of receiving your order.