Electrode Current USC
Electrode Current USC
Electrode current USC, red tip, 2 mm pin
Catalog # 77664-240
Supplier #:  EKC
Electrode Current USC
Catalog # 77664-240
Electrode Current USC
Electrode current USC, red tip, 2 mm pin
Price
$304.92
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Current (red) and voltage (black) electrodes are threaded and screw securely into the end of each Luer tipped cartridge. The Luer tip then plugs securely into the Luer openings of the chamber. The cable from each electrode terminates with a 2 mm pin which may be plugged into most voltage/current clamps.
  • Ussing chamber current electrode
  • Works with most voltage/current clamps
The miniature electrode gel cartridge is a small plastic tube with a male Luer tip identical to those at the tip of hypodermic syringes. The tube may be filled with different gel materials. Agar is commonly used, but other gel materials may also be satisfactory.
Specifications
  • Désignation:
    Electrode current USC, red tip, 2 mm pin