Engel Tenotomy Hook | Avantor
Engel Tenotomy Hook
13 cm (5.1"), Stainless steel
Catalog # 77663-434
Fournisseur #:  504021
Engel Tenotomy Hook
Catalog # 77663-434
Engel Tenotomy Hook
13 cm (5.1"), Stainless steel
Price
$92.46
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Engel tenotomy hook has double prongs with sharp tips.
Spécifications
  • Longueur:
    13 cm (5.1")
  • Matière:
    Stainless steel