Volkman Bone Curette | Avantor
Volkman Bone Curette
Volkman Bone Curette
Certificates
Volkman Bone Curette
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Volkman bone curette is a bone curette for debulking, and it has a razor-sharp cutting edge.
  • It is commonly used in orthopedic procedures
  • The knurled round handle of this curette gives you a sure grip
  • The handle gradually tapers to the tips
Removal of fibrous tissue from bone or scrape bone to remove small pieces or shape it with this curette. This cutting edge is an oval scoop at the end of the curette. This is manufactured for surgical grade stainless steel and is suitable for sterilization via autoclaving or chemical sterilant.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Matière
    Working Surface Style
    Availability
    Price
  • Volkman bone curette
    17 cm (6.75")
    Stainless steel
    Double ended
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $77.89
    $77.89
    Each
       $77.89
    Supplier Number: 503752
  • Volkman bone curette, cup width: 5 mm
    17 cm (6.75")
    Stainless steel
    Oval cutting surface
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $90.03
    $90.03
    Each
       $90.03
    Supplier Number: 503753