Halstead Mosquito Forceps, Disposable
Halstead Mosquito Forceps, Disposable
Halstead Mosquito Forceps, Disposable
Certificates
Halstead Mosquito Forceps, Disposable
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Halsted (or Halstead) Mosquito Forceps, also called mosquito hemostatic forceps or simply hemostats, are surgical instruments designed for clamping blood vessels or other small tissues to control bleeding during surgery or to provide a secure grip.
  • Forceps have long, thin, and straight or curved jaws that taper to a fine tip
  • Jaws are serrated for a firm grasp with minimal tissue trauma
  • Handles of mosquito forceps have a ratchet lock
They are so named because they resemble the slender legs of a mosquito. Life science researchers like these fine hemostats because of the secure grip and the ability to manipulate them in confined spaces.
Packaging: Forceps are sold in packages of 12.
Product Family Options
Product Information
  • Désignation
    Matière
    Longueur
    Curvature
    Availability
    Price
  • Halstead mosquito forcep, serrated, fine tips
    Stainless steel
    12.5 cm (5")
    Curved
    Log in for availability
    Log in for availability
    Pack of 12
    Pack of 12 - $67.92
    $67.92
    Pack of 12
       $67.92
    Supplier Number: 503724-12
    Specifications:
    More
    Specifications
    Cat. No.
    77663-396
  • Halstead mosquito forcep, serrated, fine tips
    Stainless steel
    12.5 cm (5")
    Straight
    Log in for availability
    Log in for availability
    Pack of 12
    Pack of 12 - $67.92
    $67.92
    Pack of 12
       $67.92
    Supplier Number: 503723-12
    Specifications:
    More
    Specifications
    Cat. No.
    77663-394