Iris Forceps, Disposable | Avantor
Iris Forceps, Disposable
Iris Forceps, Disposable
Certificates
Iris Forceps, Disposable
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
ris Forceps are a pair of surgical forceps (tweezers) often used in ophthalmic procedures. Iris Forceps are a small pair of forceps with a fine pointed tip for grasping delicate tissue with minimal trauma.
  • Narrow, serrated jaws of the surgical forceps
  • Fine tipped tweezers
  • Surgical grade stainless steel
The narrow, serrated jaws of the surgical forceps allow for fine manipulation of structures like the iris of the eye. Researchers like these fine tipped tweezers for precision work in microdissection. These forceps are 10 cm long with fine tips, and they come with straight or curved tips. The Iris Forceps are manufactures with surgical grade stainless steel.
Our Disposable Iris Forceps are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. These economy forceps are sold in packages of 12, perfect for students or for disposable use. All sales are final on disposable instrument packs.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Longueur
    Curvature
    Availability
    Price
  • Iris forcep, serrated
    Stainless steel
    10 cm (4")
    Curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $66.34
    $66.34
    Pack of 12
       $66.34
    Supplier Number: 503722-12
  • Iris forcep, serrated
    Stainless steel
    10 cm (4")
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $64.78
    $64.78
    Pack of 12
       $64.78
    Supplier Number: 503721-12