Adson Forceps, Straight
Adson Forceps, Straight
Adson Forceps, Straight
Certificates
Adson Forceps, Straight
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.
Packaging: Package of 12.
Product Family Options
Product Information
  • Désignation
    Matière
    Longueur
    Curvature
    Availability
    Price
  • Adson forcep, 1×2 teeth
    Stainless steel
    12 cm (4.75")
    Straight
    Log in for availability
    Log in for availability
    Pack of 12
    Pack of 12 - $71.08
    $71.08
    Pack of 12
       $71.08
    Supplier Number: 503720-12
  • Adson forcep, serrated
    Stainless steel
    12 cm (4.75")
    Straight
    Log in for availability
    Log in for availability
    Pack of 12
    Pack of 12 - $64.78
    $64.78
    Pack of 12
       $64.78
    Supplier Number: 503719-12