EVOM™ Auto Wired Connection Kit
EVOM™ auto wired connection kit
  77664-266
 :  EVA-AC-03-01
EVOM™ Auto Wired Connection Kit
  77664-266
EVOM™ Auto Wired Connection Kit
EVOM™ Auto Wired Connection Kit
EVOM™ auto wired connection kit
$150.10
Each
This kit is for peer-to peer wired connection set up.
The set up requires EVOM™ Auto system’s Software rev 3.0.0.
 : This kit contains:
(1) USB 3.0 GB ethernet adapter
(1) Ethernet cable 25'
(1) EVOM™ auto ethernet update USB flash drive
  • Désignation:
    EVOM™ auto wired connection kit