Zero Pressure Weather Caps, Labconco®
Zero Pressure Weather Caps, Labconco®
Supplier:   Labconco
Zero Pressure Weather Caps, Labconco®
Supplier:  Labconco
Certificates
Zero Pressure Weather Caps, Labconco®
Zero Pressure Weather Caps, Labconco®
5 Options Available Below
Product Details & Documents
Weathercaps are constructed of strong, corrosion-resistant PVC.
Caps fits atop standard PVC duct, and permit vertical discharge of effluent air above roofline for dispersion away from the building. Caps add little static pressure to the exhaust system.
Product Family Options
Informations sur le produit
  • Désignation
    Hauteur
    Nominal Diameter
    Shipping Weight
    Availability
    Price
  • Zero Pressure Weathercaps
    91.4 cm (36")
    15.2 cm (6")
    9.1 kg (20 lbs.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,380.00
    $1,380.00
    Each
       $1,380.00
    Minimum order quantity is 1
    Supplier Number: 4722200
  • Zero Pressure Weather Caps
    101.6 cm (40")
    20.3 cm (8")
    11.3 kg (25 lbs.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,700.00
    $1,700.00
    Each
       $1,700.00
    Supplier Number: 4722300
  • Zero Pressure Weather Caps
    121.9 cm (48")
    25.4 cm (10")
    13.6 kg (30 lbs.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,010.00
    $2,010.00
    Each
       $2,010.00
    Minimum order quantity is 1
    Supplier Number: 7095100
  • Zero Pressure Weather Caps
    142.2 cm (56")
    30.5 cm (12")
    15.9 kg (35 lbs.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,340.00
    $2,340.00
    Each
       $2,340.00
    Minimum order quantity is 1
    Supplier Number: 5622100
  • Zero Pressure Weather Caps
    182.9 cm (72")
    40.6 cm (16")
    18.1 kg (40 lbs.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,990.00
    $2,990.00
    Each
       $2,990.00
    Supplier Number: 5622200