ContactUsLinkComponent
Alconox® Powder Precision Cleaner
Catalogue # 77661-644
New Product
Alconox® Powder Precision Cleaner
Catalogue # 77661-644
Supplier Number:  13740
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Powder precision cleaner
  • Cat. no.:
    77661-644
  • pH:
    9.5
  • Weight:
    4 lbs.
Spécifications
À propos de cet article
Alconox® is a concentrated, anionic detergent for manual and ultrasonic cleaning. This detergent gives you reliable results without interfering residues, because it rinses away easily. Alconox cleaner is Ideal for cleaning contaminants from glassware, metals, plastics, ceramic, porcelain, rubber and fiberglass. It is an excellent replacement for corrosive acids and hazardous solvents.
  • Save money with a concentrated powder that can be diluted 1:100
  • Protect the environment with this biodegradable and readily disposable cleaning solution
  • Save time with penetrating wetting power
  • Get reliable results with no interfering residues on labware
  • Free rinsing means it rinses away easily for residue-free results
  • Corrosion inhibited means Alconox is even safe on soft metals, plastics, rubber and fiberglass
  • Use with manual or ultrasonic cleaning methods
  • Pass your cleaning validation tests for lab accreditation and plant inspection approval using USDA authorized Alconox
Alconox® can be used as a paste, as a soak or ultrasonically. It can be used on glass, metal, stainless steel, porcelain, ceramic, plastic, rubber and fiberglass. It can also be used on soft metals such as copper aluminum, zinc and magnesium, if they are rinsed promptly.
It can be used to remove: soil, grit, grime, buffing compound, slime, grease, blood, tissue, oils, salts, deposits, particulates, solvents, chemicals, radioisotopes, radioactive contaminants, silicone oils and mold release agents.