SMARTouch™ Microinjection Syringe Pump Controller | Avantor
SMARTouch™ Microinjection Syringe Pump Controller
SMARTouch™ Controller only, two-channel
Catalog # 77664-816
Fournisseur #:  MICRO2T
SMARTouch™ Microinjection Syringe Pump Controller
Catalog # 77664-816
SMARTouch™ Microinjection Syringe Pump Controller
SMARTouch™ Microinjection Syringe Pump Controller
SMARTouch™ Controller only, two-channel
Price
$3,276.92
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The touch-screen display on WPI's MICRO2T SMARTouch™ controller offers an intuitive, user-friendly interface. Our MICRO2T allows for simultaneous control of the UMP3 syringe pump.
  • Splash-proof touch screen
  • Built-in memory configured for saving settings between use
  • A dual mode motor drive
The MICRO2T SMARTouch™ controller for the UMP3 features patent pending technology which includes:
Total system calibration - Calibrate the syringe and the controller together as a system. This feature eliminates variability of the syringe and delivers (or withdraws) the configured volume.
Smart smoothness - The controller can be set to automatically adjust microstepping according to the injection rate to deliver (or withdraw) smoothly.
User defined travel limits - Set the limits for your NanoSyringe on your MICRO2T. This prevents your UMP3 from exceeding its drive range.
The MICRO2T SMARTouch™ controller has a graphical indication of the rate and volume remaining in your NanoSyringe. It offers end-stop detection that is dependent on the syringe volume. You can control two pumps independently the MICRO2T's dual capability. The controller also has automatic pump detection and a Pause/Resume feature for incremental dosing during infusion, or sample collection upon withdraw. The MICRO2T SMARTouch™ controller is fully compatible with all previous models of the UMP3.
Spécifications
  • Désignation:
    SMARTouch™ Controller only, two-channel
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Foot switch for the MICRO2T controller
    Foot switch for the MICRO2T controller
    77661-618
    Log in pour la disponibilité
    Each - $333.38