Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Spécifications
- Description:Cross-action microdissecting forceps
- Length:8.9 cm (3.5")
- Tip Configuration:Straight
- Cat. no.:77663-606
Spécifications
À propos de cet article
Cross-action tweezers or cross-action forceps, have a distinctive design that allows for automatic closure.
- Surgical forceps are made of surgical grade stainless steel
- Sterilize them in an autoclave or by chemical sterilization
These forceps have a cross-over or scissor-like mechanism in the handle, which lets you clamp tissues, reducing hand fatigue and giving you a secure, consistent hold. Cross-action dissecting forceps are perfect for fine tissue dissection, giving you a secure grip without the use of excessive pressure. The fine tips are a benefit in microsurgery when you are working in confined spaces.
They are commonly used in ophthalmic procedures, neurosurgery, and vascular surgeries.
Clean and inspect your surgical instruments frequently.