ContactUsLinkComponent
Enzol Enzymatic Detergent
Enzol Enzymatic Detergent
Catalog # 77663-938
New Product
Enzol Enzymatic Detergent
Catalog # 77663-938
Supplier Number:  7363-4
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Enzol Enzymatic detergent
  • Size:
    1 gal.
  • Cat. no.:
    77663-938
Specifications
About this item
Dirty surgical instruments with remnants of hidden debris, built-up proteins and spots of rust can corrupt you research results, invalidating all your work.
  • Fast, effective protealytic detergent designed for cleaning surgical instruments
  • High Level Disinfection (HLD), which kills most microbes and some bacterial spores
  • The CDC recommends a 90 minute soak at 25 °C
After a good cleaning, you may proceed to disinfection and sterilization according to your GLP (good laboratory practice). Cleaning instruments is always the first step towards sterilization. A detergent like Enzol assists in the removal of the debris that could also interfere with further disinfection/sterilization.