To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Enzol Enzymatic detergent
- Size:1 gal.
- Cat. no.:77663-938
Specifications
About this item
Dirty surgical instruments with remnants of hidden debris, built-up proteins and spots of rust can corrupt you research results, invalidating all your work.
- Fast, effective protealytic detergent designed for cleaning surgical instruments
- High Level Disinfection (HLD), which kills most microbes and some bacterial spores
- The CDC recommends a 90 minute soak at 25 °C
After a good cleaning, you may proceed to disinfection and sterilization according to your GLP (good laboratory practice). Cleaning instruments is always the first step towards sterilization. A detergent like Enzol assists in the removal of the debris that could also interfere with further disinfection/sterilization.