Noyes Spring Scissors, Angled Up | Avantor
Noyes Spring Scissors, Angled Up
Noyes spring scissors, angled on the side, 12 cm (4.75"), 14 mm, Sharp
Catalog # 77663-628
Fournisseur #:  504491
Noyes Spring Scissors, Angled Up
Catalog # 77663-628
Noyes Spring Scissors, Angled Up
Noyes spring scissors, angled on the side, 12 cm (4.75"), 14 mm, Sharp
Price
$262.79
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Noyes micro scissors are a surgical instrument designed for ophthalmic surgery and are an ideal choice for microdissection or for delicate small animal surgeries. Named after their inventor, ophthalmologist Dr. Arthur Noyes, the Noyes Micro Scissors are characterized by their small size and fine tips.
  • Made from premium German stainless steel, which is suitable for autoclaving or chemical sterilization
  • Spring scissors for ambidextrous use
These surgical scissors are typically used for delicate tasks such as cutting fine structures, trimming tissue, or creating small openings. For this reason, Noyes scissors are often preferred by left-handed surgeons.
Always clean your surgical instruments promptly after you use them. Proper cleaning lengthens the usable lifespan of surgical instruments.
Spécifications
  • Désignation:
    Noyes spring scissors, angled on the side
  • Longueur:
    12 cm (4.75")
  • Blade Length:
    14 mm
  • Blade Type:
    Sharp