Nalgene® Lower Assemblies/Shafts and Impellers, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
À propos de cet article
Components are for use with BioTech Mixer Overhead Drive (see 66463-212 series)
Recommended vessel (sold separately) is listed below. Lower assemblies use type 316 stainless steel axial-flow impellers and shafts. All require sanitary mixer support (see 66463-210 series).
Frequently Bought Together