Compact Roller System for Mini Bottles, Wheaton®, DWK Life Sciences
À propos de cet article
Designed for small-scale, benchtop mixing and agitation
Compact size permits use in standard incubators and cold rooms. Continuous rotation of cylindrical bottles at predetermined optimum speeds provides better distribution of media and uniform gassing. Transistorized solid state circuitry allows exact speed control of the bottles and is designed to compensate against sudden or prolonged line voltage changes. Roller shafts are mounted in self-lubricating nylon bearings, which eliminate many maintenance problems. Use in CO2 environments may necessitate more frequent replacement of motor brushes. Up to two additional deck kits can be added to accommodate an additional four bottles per kit.
For 30mL or larger serum bottles, 100–125mL media bottles, or 38x200mm culture tubes for the growth and observation of single chicken or rat embryos. Accommodates a maximum of four bottles that are 38–60mm in diameter, and up to 240mm long.
Certifications: UL listed. CSA certified. CE marked.
Ordering information: Supplied without bottles. Deck kit consists of assembled roller deck, four support posts, drive belt, necessary hardware, and assembly instructions.
2 Options Available Below
Product Details & Documents
Designed for small-scale, benchtop mixing and agitation
Compact size permits use in standard incubators and cold rooms. Continuous rotation of cylindrical bottles at predetermined optimum speeds provides better distribution of media and uniform gassing. Transistorized solid state circuitry allows exact speed control of the bottles and is designed to compensate against sudden or prolonged line voltage changes. Roller shafts are mounted in self-lubricating nylon bearings, which eliminate many maintenance problems. Use in CO2 environments may necessitate more frequent replacement of motor brushes. Up to two additional deck kits can be added to accommodate an additional four bottles per kit.
For 30mL or larger serum bottles, 100–125mL media bottles, or 38x200mm culture tubes for the growth and observation of single chicken or rat embryos. Accommodates a maximum of four bottles that are 38–60mm in diameter, and up to 240mm long.
Certifications: UL listed. CSA certified. CE marked.
Ordering information: Supplied without bottles. Deck kit consists of assembled roller deck, four support posts, drive belt, necessary hardware, and assembly instructions.
Specifications for Product Family
Spécifications
- Catalog No:
- 14231-438
- 14231-486
- Bottle speed:
- 3 - 45 rpm (38 mm dia. bottle)
2 - 30 rpm (60 mm dia. bottle) - 3 - 45 rpm (38 mm dia. bottle)
2 - 30 rpm (60 mm dia. bottle)
- Electrical:
- 100–120V, 50/60Hz, 14W
- 100–120V, 50/60Hz, 14W
- Max. ambient temperature:
- 40°C (104°F)
- 40°C (104°F)
- Shipping weight:
- 8 kg (18 lbs.)
- 8 kg (18 lbs.)
- Dimensions:
- 31W x 32D x 18H cm (121/4 x 123/4 x 71/8")
- 31W x 32D x 18H cm (121/4 x 123/4 x 71/8")
Spécifications
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Roller System for Mini Bottles, Single DeckLog in pour la disponibilitéLog in pour la disponibilitéCase of 1Case of 1 - $3,421.60$3,421.60Prix promotionnel pour les nouveaux clientsCase of 1$3,421.60Supplier Number: W348923-ASpécifications:CSA CertifiedSmall Business EnterpriseUL Listed
More
Specifications
Cat. No.14231-438Bottle speed3 - 45 rpm (38 mm dia. bottle)
2 - 30 rpm (60 mm dia. bottle)Electrical100–120V, 50/60Hz, 14WMax. ambient temperature40°C (104°F)Shipping weight8 kg (18 lbs.)Dimensions31W x 32D x 18H cm (121/4 x 123/4 x 71/8") - Deck KitLog in pour la disponibilitéLog in pour la disponibilitéCase of 1Case of 1 - $1,183.00$1,183.00Prix promotionnel pour les nouveaux clientsCase of 1$1,183.00Supplier Number: W348920-CHSpécifications:CSA CertifiedSmall Business EnterpriseUL Listed
More
Specifications
Cat. No.14231-486Bottle speed3 - 45 rpm (38 mm dia. bottle)
2 - 30 rpm (60 mm dia. bottle)Electrical100–120V, 50/60Hz, 14WMax. ambient temperature40°C (104°F)Shipping weight8 kg (18 lbs.)Dimensions31W x 32D x 18H cm (121/4 x 123/4 x 71/8")CSA CertifiedSmall Business EnterpriseUL Listed