SAF-T-FIT™ Plus N1139 Disposable Particulate Respirators, Honeywell Safety
Fournisseur: Honeywell Safety Products
Certificats
À propos de cet article
Provides 99% particulate efficiency against oil-free particulates
- Equipped with exhalation valve for ease of breathing and exceptional comfort during extended wear
- Full sealing flange for superior fit and protection
- Latex-free
Ideal for welding, soldering, metal cutting and pouring, torch cutting, grinding, sanding, sweeping, hot temperatures, general industrial applications, and dusty operations.
Frequently Bought Together