8212 Disposable N95 Particulate Respirators, 3M™
Catalogue # CA56621-926
Fournisseur: 3M
Spécifications
- Description:8212 N95 Respirator
- Cat. no.:CA56621-926
- Supplier No.:70070708998
Spécifications
À propos de cet article
Designed for use when welding, torch cutting, soldering, brazing and working with molten metals
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
Helps provide protection when welding on galvanized and stainless steel. Fits easily under a welding hood. The respirator filter is at least 95% efficient at removing solid and liquid particles (excluding those containing oil). Adjustable straps, face seal, and Cool Flow® exhalation valve provide added comfort and durability. Caking-resistant filter media construction allows for easier breathing over long periods of wear.