ContactUsLinkComponent
8210 Disposable N95 Respirator, 3M™
Catalog # CA56222-362
Supplier:  3M
undefined
8210 Disposable N95 Respirator, 3M™
Catalog # CA56222-362
Supplier:  3M
Supplier Number:  7100132742
Specifications
  • Description:
    8210 N95 Respirator
  • Cat. no.:
    CA56222-362
  • Supplier No.:
    8210
Specifications
About this item
Economical respirator provides reliable worker protection against certain solid and liquid aerosols that do not contain oil.
  • Provides reliable worker protection against certain solid and liquid aerosols that do not contain oil
  • Provides minimum filtration efficiency of 95%
  • Lightweight construction for greater worker comfort
  • Adjustable nose clip for custom fit and secure seal
Provides minimum filtration efficiency of 95%. Lightweight construction for greater worker comfort. Adjustable noseclip for custom fit and secure seal.
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.