Synthware Buchner Filter Funnels, Inner [ST] Joint, Kemtech America
These filter funnels are made of glass.
- Heavy wall borosilicate construction
- Ground glass joint
- Medium and coarse frit porosity available
- Barbed side arm
Note: Not recommended for filtration where pressure would exceed 15 psi.
- Catalog No:
- 60003-484
- 60003-486
- 60003-452
- 60003-454
- 60003-456
- 60003-458
- 60003-488
- 60003-444
- 60003-490
- 60003-446
- 60003-492
- 60003-448
- 60003-494
- 60003-450
- 60003-502
- 60003-460
- 60003-462
- 60003-464
- 60003-500
- 60003-474
- 60003-476
- 60003-478
- 60003-466
- 60003-468
- 60003-470
- 60003-472
- Hose connection ext.Ø:
- 8 mm
- 8 mm
- 8 mm
- 8 mm
- 8 mm
- 8 mm
- 8 mm
- 8 mm
- 8 mm
- 8 mm
- 8 mm
- 8 mm
- 8 mm
- 8 mm
- 8 mm
- 8 mm
- 8 mm
- 8 mm
- 8 mm
- 8 mm
- 8 mm
- 8 mm
- 8 mm
- 8 mm
- 8 mm
- 8 mm
Frequently Bought Together