KIMTECH PURE® M7 Face Veil With Knitted Headbands, KIMBERLY-CLARK PROFESSIONAL®
Fournisseur: Microflex
Certificats
À propos de cet article
Single-use personal veils are designed to be used as a hygienic alternative to reusable garments.
- Low-linting apertured polyethylene film, filters and minimizes contamination risks
- Exceptional breathability
- Headband versions usable with reusable or disposable garment systems
Recommended for ISO Class 7 or higher cleanroom environments.
A covered nose piece allows veils to conform to the nose and cheekbones, ensuring a secure and comfortable fit. Veils are available with snaps or knitted headband. Veils with snaps will fit into most reusable hoods.
Ordering information: All veils are supplied double bagged.
Frequently Bought Together